Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P3H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001529.1 | NP_001001529.1 | 3 - 4 | 786 | 77 | |
GenBank mRNA | AY463528.1 | - | 3 - 4 | 786 | 77 | |
BX931522.1 | - | 3 - 4 | 809 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |