Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC24A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001773.1 | NP_001001773.1 | 2 - 3 | 1049 | 57 | |
GenBank mRNA | AF177984.1 | - | 2 - 3 | 1049 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |