Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004375.2 | NP_001004375.1 | 2 - 3 | 328 | 67 | |
GenBank mRNA | BX274578.4 | - | 2 - 3 | 334 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |