Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004765.2 | NP_001004765.2 | 2 - 3 | 492 | 87 | |
NM_001317827.1 | NP_001304756.1 | 2 - 3 | 492 | 87 | ||
NM_001364409.1 | NP_001351338.1 | 2 - 3 | 492 | 87 | ||
GenBank mRNA | AB190358.1 | - | 2 - 3 | 493 | 87 | |
AJ829442.1 | - | 1 - 2 | 169 | 87 | ||
BX929627.2 | - | 2 - 3 | 468 | 87 | ||
CN383036.1 | - | 1 - 2 | 112 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |