Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AXIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204944.2 | NP_990275.1 | 9 - 10 | 2652 | 70 | |
XM_015294136.2 | XP_015149622.1 | 9 - 10 | 2882 | 70 | ||
XM_015294137.2 | XP_015149623.1 | 8 - 9 | 2708 | 70 | ||
XM_025155049.1 | XP_025010817.1 | 10 - 11 | 2740 | 70 | ||
XM_025155050.1 | XP_025010818.1 | 10 - 11 | 2737 | 70 | ||
GenBank mRNA | AF009012.1 | - | 9 - 10 | 2652 | 70 | |
AY640375.1 | - | 9 - 10 | 2548 | 70 | ||
AY687628.1 | - | 8 - 9 | 2395 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |