Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205026.1 | NP_990357.1 | 13 - 14 | 1179 | 82 | |
XM_015296335.2 | XP_015151821.1 | 13 - 14 | 1196 | 82 | ||
XM_025142176.1 | XP_024997944.1 | 13 - 14 | 1203 | 82 | ||
GenBank mRNA | AB219157.1 | - | 12 - 13 | 1130 | 82 | |
AB219158.1 | - | 12 - 13 | 1130 | 82 | ||
AB219159.1 | - | 12 - 13 | 1130 | 82 | ||
AB219160.1 | - | 12 - 13 | 1130 | 82 | ||
AF029841.1 | - | 13 - 14 | 1179 | 82 | ||
CR353912.1 | - | 5 - 6 | 359 | 82 | ||
HG007958.1 | - | 13 - 14 | 1209 | 82 | ||
LC144559.1 | - | 12 - 13 | 1130 | 82 | ||
LC144560.1 | - | 12 - 13 | 1130 | 82 | ||
LC144561.1 | - | 12 - 13 | 1130 | 82 | ||
LC144562.1 | - | 12 - 13 | 1130 | 82 | ||
LC144563.1 | - | 12 - 13 | 1130 | 82 | ||
LC144564.1 | - | 12 - 13 | 1130 | 82 | ||
LC144565.1 | - | 12 - 13 | 1130 | 82 | ||
LC144566.1 | - | 12 - 13 | 1130 | 82 | ||
LC144567.1 | - | 12 - 13 | 1130 | 82 | ||
LC144568.1 | - | 12 - 13 | 1130 | 82 | ||
LC144569.1 | - | 12 - 13 | 1130 | 82 | ||
LC144571.1 | - | 12 - 13 | 1130 | 82 | ||
LC144572.1 | - | 12 - 13 | 1130 | 82 | ||
LC144573.1 | - | 12 - 13 | 1130 | 82 | ||
LC144574.1 | - | 12 - 13 | 1130 | 82 | ||
LC144575.1 | - | 12 - 13 | 1130 | 82 | ||
LC144576.1 | - | 12 - 13 | 1130 | 82 | ||
LC144578.1 | - | 12 - 13 | 1130 | 82 | ||
LC144579.1 | - | 12 - 13 | 1130 | 82 | ||
LC144580.1 | - | 12 - 13 | 1130 | 82 | ||
LC144582.1 | - | 12 - 13 | 1130 | 82 | ||
LC144583.1 | - | 12 - 13 | 1130 | 82 | ||
LC144584.1 | - | 12 - 13 | 1130 | 82 | ||
LC144585.1 | - | 12 - 13 | 1130 | 82 | ||
LC144586.1 | - | 12 - 13 | 1130 | 82 | ||
LC144587.1 | - | 12 - 13 | 1130 | 82 | ||
LC144588.1 | - | 12 - 13 | 1130 | 82 | ||
LC144589.1 | - | 12 - 13 | 1130 | 82 | ||
LC144590.1 | - | 12 - 13 | 1130 | 82 | ||
LC144591.1 | - | 12 - 13 | 1130 | 82 | ||
LC144592.1 | - | 12 - 13 | 1130 | 82 | ||
LC144593.1 | - | 12 - 13 | 1130 | 82 | ||
LC144594.1 | - | 12 - 13 | 1130 | 82 | ||
LC144595.1 | - | 12 - 13 | 1130 | 82 | ||
LC144596.1 | - | 12 - 13 | 1130 | 82 | ||
LC144597.1 | - | 12 - 13 | 1130 | 82 | ||
LC144598.1 | - | 12 - 13 | 1130 | 82 | ||
LC144599.1 | - | 12 - 13 | 1130 | 82 | ||
LC144600.1 | - | 12 - 13 | 1130 | 82 | ||
LC144601.1 | - | 12 - 13 | 1130 | 82 | ||
LC144602.1 | - | 12 - 13 | 1130 | 82 | ||
LC144603.1 | - | 12 - 13 | 1130 | 82 | ||
LC144604.1 | - | 12 - 13 | 1130 | 82 | ||
LC144605.1 | - | 12 - 13 | 1130 | 82 | ||
LC144606.1 | - | 12 - 13 | 1130 | 82 | ||
LC144607.1 | - | 12 - 13 | 1130 | 82 | ||
LC144608.1 | - | 12 - 13 | 1130 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |