Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031247.1 | NP_001026418.1 | 1 - 2 | 127 | 48 | |
XM_015289173.2 | XP_015144659.1 | 1 - 2 | 159 | 48 | ||
XM_015289174.2 | XP_015144660.1 | 1 - 2 | 125 | 48 | ||
GenBank mRNA | AJ720729.1 | - | 1 - 2 | 127 | 48 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |