Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031468.1 | NP_001026639.1 | 5 - 6 | 480 | 137 | |
GenBank mRNA | AJ720584.1 | - | 5 - 6 | 480 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |