Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030654.2 | NP_001025825.2 | 5 - 6 | 408 | 89 | |
XM_015275241.2 | XP_015130727.1 | 4 - 5 | 596 | 89 | ||
XM_025155434.1 | XP_025011202.1 | 3 - 4 | 629 | 89 | ||
XM_025155435.1 | XP_025011203.1 | 4 - 5 | 3598 | 89 | ||
XM_025155436.1 | XP_025011204.1 | 5 - 6 | 4167 | 89 | ||
XM_025155437.1 | XP_025011205.1 | 4 - 5 | 546 | 89 | ||
GenBank mRNA | AJ720671.1 | - | 5 - 6 | 413 | 89 | |
AJ851383.1 | - | 5 - 6 | 408 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |