Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030775.1 | NP_001025946.1 | 4 - 5 | 636 | 81 | |
GenBank mRNA | AJ720533.1 | - | 4 - 5 | 636 | 81 | |
CR385271.1 | - | 4 - 5 | 506 | 81 | ||
CR389600.1 | - | 4 - 5 | 467 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |