Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205328.5 | NP_990659.1 | 2 - 3 | 463 | 67 | |
XM_015293745.2 | XP_015149231.1 | 3 - 4 | 3609 | 67 | ||
XM_015293746.2 | XP_015149232.1 | 3 - 4 | 3609 | 67 | ||
XM_015293747.2 | XP_015149233.1 | 3 - 4 | 3609 | 67 | ||
XM_015293748.2 | XP_015149234.1 | 3 - 4 | 3609 | 67 | ||
XM_015293750.2 | XP_015149236.1 | 3 - 4 | 3609 | 67 | ||
XM_015293751.2 | XP_015149237.1 | 3 - 4 | 3609 | 67 | ||
XM_015293752.2 | XP_015149238.1 | 3 - 4 | 3609 | 67 | ||
GenBank mRNA | CR352392.1 | - | 3 - 4 | 361 | 67 | |
U14942.1 | - | 2 - 3 | 410 | 67 | ||
X80340.1 | - | 467 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |