Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077233.1 | NP_001070701.1 | 23 - 24 | 2972 | 62 | |
GenBank mRNA | DQ224386.1 | - | 23 - 24 | 2972 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |