Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204213.1 | NP_989544.1 | 1 - 2 | 194 | 92 | |
XM_015292118.2 | XP_015147604.1 | 2 - 3 | 328 | 92 | ||
XM_025149632.1 | XP_025005400.1 | 2 - 3 | 328 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |