Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204288.1 | NP_989619.1 | 4 - 5 | 659 | 64 | |
XM_015292934.2 | XP_015148420.1 | 4 - 5 | 642 | 64 | ||
XM_015292935.1 | XP_015148421.1 | 4 - 5 | 642 | 64 | ||
XM_025154402.1 | XP_025010170.1 | 4 - 5 | 642 | 64 | ||
XM_025154403.1 | XP_025010171.1 | 4 - 5 | 695 | 64 | ||
GenBank mRNA | AF439559.1 | - | 4 - 5 | 659 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |