Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204418.1 | NP_989749.1 | 3 - 4 | 504 | 73 | |
GenBank mRNA | L08047.1 | - | 3 - 4 | 504 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |