Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204460.1 | NP_989791.1 | 2 - 3 | 336 | 79 | |
GenBank mRNA | AJ493595.1 | - | 2 - 3 | 336 | 79 | |
AM773756.1 | - | 2 - 3 | 267 | 79 | ||
AY744450.1 | - | 2 - 3 | 273 | 79 | ||
AY920750.1 | - | 2 - 3 | 274 | 79 | ||
FJ788635.1 | - | 1 - 2 | 273 | 79 | ||
HM747027.1 | - | 2 - 3 | 273 | 79 | ||
HQ008777.1 | - | 2 - 3 | 273 | 79 | ||
HQ340168.1 | - | 2 - 3 | 273 | 79 | ||
HQ340169.1 | - | 2 - 3 | 273 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |