Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204525.1 | NP_989856.1 | 2 - 3 | 323 | 67 | |
XM_015299568.2 | XP_015155054.1 | 3 - 4 | 462 | 67 | ||
GenBank mRNA | Y07811.1 | - | 323 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |