Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005825.1 | NP_001005825.1 | 3 - 4 | 394 | 134 | |
XM_015292093.2 | XP_015147579.1 | 3 - 4 | 471 | 134 | ||
GenBank mRNA | AJ720263.1 | - | 3 - 4 | 394 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |