Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006499.1 | NP_001006499.1 | 5 - 6 | 594 | 72 | |
GenBank mRNA | AJ720604.1 | - | 5 - 6 | 594 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |