Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006521.2 | NP_001006521.2 | 2 - 3 | 275 | 82 | |
XM_015289800.2 | XP_015145286.1 | 2 - 3 | 289 | 82 | ||
GenBank mRNA | AJ720224.1 | - | 2 - 3 | 275 | 82 | |
CR524416.1 | - | 2 - 3 | 235 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |