Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014319.hg.1 |
TC01000909.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014319.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014319.hg.1 |
ENST00000424264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014319.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
ENST00000475289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
ENST00000475289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
ENST00000475289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014309.hg.1 |
TC01000909.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014309.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014314.hg.1 |
TC01000909.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014314.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014314.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014317.hg.1 |
TC01000909.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014317.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014317.hg.1 |
ENST00000424264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014317.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014317.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014318.hg.1 |
TC01000909.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014318.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014318.hg.1 |
ENST00000424264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014318.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014318.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014313.hg.1 |
TC01000909.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014313.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014313.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014316.hg.1 |
TC01000909.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014316.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014316.hg.1 |
ENST00000424264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014316.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014316.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014312.hg.1 |
TC01000909.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014312.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014312.hg.1 |
ENST00000424264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014312.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014312.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165831.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165831.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165831.hg.1 |
TC0100009241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100165837.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165837.hg.1 |
ENST00000475289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165837.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165837.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165837.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165837.hg.1 |
ENST00000475289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165837.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165837.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165837.hg.1 |
TC0100009241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100165840.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165840.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165840.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165840.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165840.hg.1 |
TC0100009241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100165835.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165835.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165835.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165835.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165835.hg.1 |
TC0100009241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
ENST00000475289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
ENST00000475289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
ENST00000475289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009241.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165834.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165834.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165834.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165834.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165834.hg.1 |
TC0100009241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100165838.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165838.hg.1 |
ENST00000475289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165838.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165838.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165838.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165838.hg.1 |
ENST00000475289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165838.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165838.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165838.hg.1 |
TC0100009241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100165832.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165832.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165832.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165832.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165832.hg.1 |
TC0100009241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100165833.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165833.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165833.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165833.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165833.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165833.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165833.hg.1 |
TC0100009241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100165839.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165839.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165839.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165839.hg.1 |
ENST00000305352.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165839.hg.1 |
ENST00000475821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165839.hg.1 |
NM_001400.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165839.hg.1 |
TC0100009241.hg.1 |
Clariom D TC |