Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000265538.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000265538.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000265538.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000431929.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000431929.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000431929.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000431929.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000431929.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000431929.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000445425.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000445425.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000445425.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000445425.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000445425.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000445425.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000454011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000454011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000454011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000454011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000454011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000454011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
ENST00000454011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181923.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181923.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181923.hg.1 |
ENST00000431929.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181923.hg.1 |
ENST00000445425.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181923.hg.1 |
ENST00000454011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181923.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181923.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181923.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181923.hg.1 |
ENST00000431929.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181923.hg.1 |
ENST00000445425.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181923.hg.1 |
ENST00000454011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181923.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181923.hg.1 |
TC0300011060.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000265538.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000265538.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000265538.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000431929.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000431929.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000431929.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000431929.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000431929.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000431929.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000445425.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000445425.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000445425.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000445425.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000445425.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000445425.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000454011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000454011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000454011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000454011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000454011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000454011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
ENST00000454011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011060.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181921.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181921.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181921.hg.1 |
ENST00000445425.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181921.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181921.hg.1 |
ENST00000418115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181921.hg.1 |
ENST00000422781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181921.hg.1 |
ENST00000445425.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181921.hg.1 |
NM_001664.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181921.hg.1 |
TC0300011060.hg.1 |
Clariom D TC |