Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001277.hg.1 |
ENST00000262971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001277.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001277.hg.1 |
TC19000080.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000593518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000593518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000593518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000596144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000596144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000596144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000596144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000596144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000599999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000599999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000599999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000599999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000600566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000600566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000600566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000600566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000600566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000601439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000601439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000601439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
ENST00000601439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001276.hg.1 |
ENST00000262971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001276.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001276.hg.1 |
TC19000080.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900146676.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146676.hg.1 |
TC1900006670.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900146676.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146683.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146683.hg.1 |
ENST00000593518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146683.hg.1 |
ENST00000599999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146683.hg.1 |
ENST00000600566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146683.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146683.hg.1 |
TC1900006670.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900146683.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146683.hg.1 |
ENST00000593518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146683.hg.1 |
ENST00000599999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146683.hg.1 |
ENST00000600566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146683.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000593518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000593518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000593518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000596144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000596144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000596144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000596144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000596144.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000599999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000599999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000599999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000599999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000600566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000600566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000600566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000600566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000600566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000601439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000601439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000601439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
ENST00000601439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006670.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146677.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146677.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146677.hg.1 |
TC1900006670.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900146677.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146677.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146684.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146684.hg.1 |
ENST00000593518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146684.hg.1 |
ENST00000599999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146684.hg.1 |
ENST00000600566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146684.hg.1 |
NM_015897.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146684.hg.1 |
TC1900006670.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900146684.hg.1 |
ENST00000262971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146684.hg.1 |
ENST00000593518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146684.hg.1 |
ENST00000599999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146684.hg.1 |
ENST00000600566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146684.hg.1 |
NM_015897.3 |
Refseq |