Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006745.hg.1 |
ENST00000447052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011995.hg.1 |
ENST00000613436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011995.hg.1 |
ENST00000613436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000309268.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000316292.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000331523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000356303.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000356303.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000356303.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000356303.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000356303.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000356303.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000356303.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000455918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000455918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000455918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000455918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000455918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000488500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000490569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000491404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000491404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000491404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000491404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000491404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000491404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000491404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000491404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000495333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000610520.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
ENST00000615060.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012296.hg.1 |
NM_001402.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006745.hg.1 |
ENST00000447052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011995.hg.1 |
ENST00000613436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011995.hg.1 |
ENST00000613436.1 |
Ensembl |