Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001324504.1 | NP_001311433.1 | 8 | 780 | 61 | |
NM_001324505.1 | NP_001311434.1 | 6 | 748 | 61 | ||
NM_001324506.1 | NP_001311435.1 | 5 | 654 | 61 | ||
NM_020382.4 | NP_065115.3 | 7 | 870 | 61 | ||
NR_136910.1 | 8 | 983 | 61 | |||
XM_006719394.3 | XP_006719457.1 | 6 | 700 | 61 | ||
XM_011538333.2 | XP_011536635.1 | 6 | 1086 | 61 | ||
GenBank mRNA | AF287261.2 | - | 1 | 1060 | 61 | |
AK292645.1 | - | 7 | 838 | 61 | ||
AY064546.1 | - | 1 | 922 | 61 | ||
AY102937.1 | - | 7 | 800 | 61 | ||
BC050346.1 | - | 7 | 1213 | 4838030 | 61 | |
HQ259683.1 | - | 7 | 697 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon |