Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054048.hg.1 |
TC01003474.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054048.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054048.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054048.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054049.hg.1 |
TC01003474.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054049.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054049.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054049.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100223915.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100223915.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100223915.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100223915.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100223915.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100223915.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100223915.hg.1 |
TC0100016290.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100223914.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100223914.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100223914.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100223914.hg.1 |
ENST00000367874.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100223914.hg.1 |
ENST00000467021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100223914.hg.1 |
NM_053053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100223914.hg.1 |
TC0100016290.hg.1 |
Clariom D TC |