Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276727.1 | NP_001263656.1 | 1 - 2 | 93 | 81 | |
NM_138787.3 | NP_620142.2 | 1 - 2 | 93 | 81 | ||
GenBank mRNA | AK290833.1 | - | 1 - 2 | 86 | 81 | |
BC009561.1 | - | 1 - 2 | 67 | 3900485 | 81 | |
DQ121386.1 | - | 1 - 2 | 75 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |