Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000586724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000619274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000619274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
ENST00000619274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16002128.hg.1 |
ENST00000299320.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16002128.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16002128.hg.1 |
TC16000135.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16002129.hg.1 |
ENST00000299320.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16002129.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16002129.hg.1 |
TC16000135.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600148215.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148215.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148215.hg.1 |
ENST00000586724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148215.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148215.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600148215.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148215.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148215.hg.1 |
ENST00000586724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148215.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148215.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600148215.hg.1 |
TC1600006754.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000586724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000619274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000619274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
ENST00000619274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006754.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600148217.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148217.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148217.hg.1 |
ENST00000586724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148217.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148217.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600148217.hg.1 |
ENST00000299320.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148217.hg.1 |
ENST00000586256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148217.hg.1 |
ENST00000586724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148217.hg.1 |
ENST00000590191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600148217.hg.1 |
NM_139170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600148217.hg.1 |
TC1600006754.hg.1 |
Clariom D TC |