Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07016580.hg.1 |
TC07001080.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07016580.hg.1 |
ENST00000316495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07016580.hg.1 |
ENST00000401903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07016580.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000471448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000471448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000471448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000471448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000471448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000478137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000478137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000478137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000478137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000484977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000484977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000484977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000484977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000484977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000484977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000484977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000574135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000574135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000574135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
ENST00000574135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011967.hg.1 |
ENST00000498515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000471448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000471448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000471448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000471448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000471448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000478137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000478137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000478137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000478137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000484977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000484977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000484977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000484977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000484977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000484977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000484977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000574135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000574135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000574135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
ENST00000574135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010005.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011967.hg.1 |
ENST00000498515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700171173.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700171173.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700171173.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700171173.hg.1 |
ENST00000471448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700171173.hg.1 |
ENST00000574135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700171173.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700171173.hg.1 |
ENST00000316495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700171173.hg.1 |
ENST00000397083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700171173.hg.1 |
ENST00000401903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700171173.hg.1 |
ENST00000471448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700171173.hg.1 |
ENST00000574135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700171173.hg.1 |
NM_182491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700171173.hg.1 |
TC0700010005.hg.1 |
Clariom D TC |