Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162499.1 | NP_001155971.1 | 12 - 13 | 3261 | 61 | |
NM_012298.2 | NP_036430.1 | 10 - 11 | 2910 | 61 | ||
XM_011533503.2 | XP_011531805.1 | 12 - 13 | 3279 | 61 | ||
XM_011533504.2 | XP_011531806.1 | 12 - 13 | 3375 | 61 | ||
GenBank mRNA | AB014567.1 | - | 10 - 11 | 2838 | 61 | |
BC136592.1 | - | 12 - 13 | 3200 | 9020582 | 61 | |
BX648800.1 | - | 3 - 4 | 414 | 61 | ||
GU727621.1 | - | 12 - 13 | 3261 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons |