Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099456.2 | NP_001092926.2 | 1 - 2 | 796 | 107 | |
GenBank mRNA | AB084276.1 | - | 1 - 2 | 576 | 107 | |
AL365519.1 | - | 1 - 2 | 116 | 107 | ||
BC150534.1 | - | 1 - 2 | 300 | 107 | ||
BM985063.1 | - | 1 - 2 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30397461 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09888859 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09888861 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30397459 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09888853 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09888857 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30397469 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09888858 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30397480 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09888850 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09888849 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09888852 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30397471 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09888851 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09888847 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09888860 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09888845 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P352799 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 42867 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 10177 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P352799 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P352799 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888849 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30397469 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888845 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888859 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30397471 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888857 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888847 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30397461 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888852 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30397480 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888860 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888851 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888858 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888844 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888861 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888850 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30397471 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888857 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888851 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888852 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888859 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888849 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30397461 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888844 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888858 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888845 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888860 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888861 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888850 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30397480 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30397469 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09888847 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P352799 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16841667 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888886 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888890 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888851 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888888 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888858 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888843 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888847 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16841654 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16841616 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888897 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888906 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888854 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16841649 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888860 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888903 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16841642 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888849 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888877 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16841651 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16841619 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888880 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888840 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16841665 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888887 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888900 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16841672 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888899 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888905 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888889 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888859 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16841618 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888901 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16841626 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16841624 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888884 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16841659 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888894 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888882 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888841 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16841662 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16841650 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888852 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888893 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09888861 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001099456_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7992461 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7992462 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7992460 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 243110_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11735531_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11735531_x_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR16001183.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16001183.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16001183.hg.1 | TC16000057.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16001185.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16001185.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16001185.hg.1 | TC16000057.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16001184.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16001184.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16001184.hg.1 | TC16000057.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600011328.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011328.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011328.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011328.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011328.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011328.hg.1 | ENST00000566435.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011328.hg.1 | ENST00000566435.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011328.hg.1 | ENST00000566435.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011328.hg.1 | ENST00000566435.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011328.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011328.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011328.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011328.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011328.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16001182.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16001182.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16001182.hg.1 | TC16000057.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1600188804.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188804.hg.1 | ENST00000566435.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188804.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600188804.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188804.hg.1 | ENST00000566435.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188804.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600188804.hg.1 | TC1600011328.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600188798.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188798.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600188798.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188798.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600188798.hg.1 | TC1600011328.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600188801.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188801.hg.1 | ENST00000566435.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188801.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600188801.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188801.hg.1 | ENST00000566435.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188801.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600188801.hg.1 | TC1600011328.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600188799.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188799.hg.1 | ENST00000566435.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188799.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600188799.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188799.hg.1 | ENST00000566435.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188799.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600188799.hg.1 | TC1600011328.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600011328.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011328.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011328.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011328.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011328.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011328.hg.1 | ENST00000566435.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011328.hg.1 | ENST00000566435.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011328.hg.1 | ENST00000566435.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011328.hg.1 | ENST00000566435.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011328.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011328.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011328.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011328.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011328.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600188802.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188802.hg.1 | ENST00000566435.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188802.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600188802.hg.1 | ENST00000329610.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188802.hg.1 | ENST00000566435.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600188802.hg.1 | NM_001099456.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600188802.hg.1 | TC1600011328.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 243110_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 243110_x_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243110_x_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243110_x_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243110_x_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243110_x_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243110_x_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243110_x_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243110_x_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243110_x_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243110_x_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243110_x_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243110_x_at7 | - | - |
Human Genome U95 Set | 72948_r_at | - | - |
Human Genome U95 Set | 72948_PM_r_at | - | - |
Human Genome U133 Set | 243110_x_at | - | - |
Human Genome U133 Set | 243110_PM_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054181 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054181 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 164302 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3246678 | BM985063.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3246678 | NM_001099456.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3246678 | BM985063.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3246678 | NM_001099456.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3246678 | BM985063.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3246678 | NM_001099456.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3246678 | BM985063.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3246678 | NM_001099456.2 | human_transcriptome |