Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005759.hg.1 |
TC06000529.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005759.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005759.hg.1 |
ENST00000536757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005759.hg.1 |
ENST00000539035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005759.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005759.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000491324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000491324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000491324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005756.hg.1 |
TC06000529.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005756.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005756.hg.1 |
ENST00000536757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005756.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005760.hg.1 |
TC06000529.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005760.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005760.hg.1 |
ENST00000536757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005760.hg.1 |
ENST00000539035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005760.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005760.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005757.hg.1 |
TC06000529.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005757.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005757.hg.1 |
ENST00000536757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005757.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005757.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005754.hg.1 |
TC06000529.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005754.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154536.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154536.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154536.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154536.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154536.hg.1 |
TC0600007854.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000491324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000491324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000491324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007854.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154537.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154537.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154537.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154537.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154537.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154537.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154537.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154537.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154537.hg.1 |
TC0600007854.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600154540.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154540.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154540.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154540.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154540.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154540.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154540.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154540.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154540.hg.1 |
TC0600007854.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600154535.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154535.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154535.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154535.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154535.hg.1 |
TC0600007854.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600154539.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154539.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154539.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154539.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154539.hg.1 |
ENST00000373674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154539.hg.1 |
ENST00000611814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154539.hg.1 |
NM_001199159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154539.hg.1 |
NM_153370.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154539.hg.1 |
TC0600007854.hg.1 |
Clariom D TC |