Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102724450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125997.1 | 2 - 3 | 239 | 64 | ||
GenBank mRNA | AI473719.1 | - | 1 - 2 | 64 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ribosomal protein L22 | RPL22 |
EAP, HBP15, HBP15/L22, L22 |
-5.2kb | Sense | Chr. 1 - 6185020 - 6199619 on NCBI Build GRCh38 |
Downstream | ring finger protein 207 | RNF207 | C1orf188 | 362.0 | Anti-sense | Chr. 1 - 6205475 - 6221299 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Probe spans exons |