Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004889.hg.1 |
ENST00000225840.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004889.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004889.hg.1 |
NM_005623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004889.hg.1 |
TC17000386.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007560.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007560.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007560.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007560.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007560.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007560.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007560.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007560.hg.1 |
NM_005623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007560.hg.1 |
NM_005623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007560.hg.1 |
NM_005623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007560.hg.1 |
NM_005623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007560.hg.1 |
NM_005623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007560.hg.1 |
NM_005623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007560.hg.1 |
NM_005623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700154406.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154406.hg.1 |
NM_005623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700154406.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700154406.hg.1 |
NM_005623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700154406.hg.1 |
TC1700007560.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700007560.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007560.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007560.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007560.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007560.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007560.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007560.hg.1 |
ENST00000394620.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007560.hg.1 |
NM_005623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007560.hg.1 |
NM_005623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007560.hg.1 |
NM_005623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007560.hg.1 |
NM_005623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007560.hg.1 |
NM_005623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007560.hg.1 |
NM_005623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007560.hg.1 |
NM_005623.2 |
Refseq |