Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000319555.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000319555.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000319555.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000319555.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000319555.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000319555.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000319555.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000319555.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000319555.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000319555.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000319555.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000319555.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000319555.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000562417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000562417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000562417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000562417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000562437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000562437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000562437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000562437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000562437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000563351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000563351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000563351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000563351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000563351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000563351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000563351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000563351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000569086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000569086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000569086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000569086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000569086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
ENST00000569086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
NM_153813.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
NM_153813.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
NM_153813.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
NM_153813.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
NM_153813.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
NM_153813.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
NM_153813.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
NM_153813.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
NM_153813.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
NM_153813.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008822.hg.1 |
NM_153813.2 |
Refseq |