Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005713.hg.1 |
TC06000526.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005713.hg.1 |
ENST00000312917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005713.hg.1 |
ENST00000388715.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005713.hg.1 |
ENST00000394571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005713.hg.1 |
ENST00000457797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005713.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005713.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005713.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007833.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154329.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154329.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154329.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154329.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154329.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154329.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154329.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154329.hg.1 |
ENST00000312917.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154329.hg.1 |
ENST00000388715.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154329.hg.1 |
ENST00000394571.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154329.hg.1 |
ENST00000457797.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154329.hg.1 |
NM_001145716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154329.hg.1 |
NM_001145717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154329.hg.1 |
NM_173676.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154329.hg.1 |
TC0600007833.hg.1 |
Clariom D TC |