Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016894.hg.1 |
TC08001312.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016894.hg.1 |
ENST00000262213.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016894.hg.1 |
ENST00000521425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016894.hg.1 |
ENST00000536748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016894.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016895.hg.1 |
TC08001312.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016895.hg.1 |
ENST00000262213.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016895.hg.1 |
ENST00000521425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016895.hg.1 |
ENST00000536748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016895.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000518678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000518678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000518678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000518678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000518678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000518678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000520700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000520700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000520700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000520700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000520700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000520700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173054.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173054.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173054.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173054.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173054.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173054.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173054.hg.1 |
TC0800010765.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000518678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000518678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000518678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000518678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000518678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000518678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000520700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000520700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000520700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000520700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000520700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000520700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010765.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173056.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173056.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173056.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173056.hg.1 |
ENST00000262213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173056.hg.1 |
ENST00000521425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173056.hg.1 |
NM_014294.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173056.hg.1 |
TC0800010765.hg.1 |
Clariom D TC |