Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015985.hg.1 |
TC04001165.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015985.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015985.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015985.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015985.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000509316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000509316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000509316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000509316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000509316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000509316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000509316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000509316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000509316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000509316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010602.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172249.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172249.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172249.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172249.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172249.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172249.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172249.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172249.hg.1 |
ENST00000264318.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172249.hg.1 |
ENST00000502874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172249.hg.1 |
ENST00000508560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172249.hg.1 |
ENST00000511523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172249.hg.1 |
NM_000809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172249.hg.1 |
NM_001204266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172249.hg.1 |
NM_001204267.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172249.hg.1 |
TC0400010602.hg.1 |
Clariom D TC |