Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
ENST00000260733.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
ENST00000334433.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
ENST00000340076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
ENST00000358010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
ENST00000360561.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
ENST00000368999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
ENST00000369000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
ENST00000369001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
ENST00000369004.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
ENST00000369005.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
ENST00000453444.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
ENST00000513429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
ENST00000515273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
ENST00000515603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
NM_006997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
NM_206860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
NM_206861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
NM_206862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010731.hg.1 |
TC10000883.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000260733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000334433.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000358010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000360561.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000368997.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000368999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000369000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000369001.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000369004.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000369005.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000453444.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000490979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000513429.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000515273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000515603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
NM_001291876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
NM_001291877.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
NM_001291878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
NM_001291879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
NM_006997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
NM_206860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
NM_206861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
NM_206862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000260733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000334433.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000358010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000360561.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000368997.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000368999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000369000.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000369001.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000369004.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000369005.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000453444.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000490979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000513429.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000515273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
ENST00000515603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
NM_001291876.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
NM_001291877.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
NM_001291878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
NM_001291879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
NM_006997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
NM_206860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
NM_206861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
NM_206862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162597.hg.1 |
TC1000009140.hg.1 |
Clariom D TC |