Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013087.hg.1 |
TC0X000981.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013087.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013087.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013087.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009498.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164877.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164877.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164877.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164877.hg.1 |
ENST00000378045.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164877.hg.1 |
ENST00000483115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164877.hg.1 |
NM_173794.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164877.hg.1 |
TC0X00009498.hg.1 |
Clariom D TC |