Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025501.hg.1 |
TC01001645.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025501.hg.1 |
ENST00000235480.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025501.hg.1 |
ENST00000236914.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025501.hg.1 |
ENST00000367362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025501.hg.1 |
ENST00000542116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025501.hg.1 |
ENST00000544748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025501.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025501.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367357.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367357.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367357.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367357.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000447034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000447034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000447034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000474307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000474307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000474307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000474307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000474307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000474307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000474307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367357.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367357.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367357.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367357.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000447034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000447034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000447034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000474307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000474307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000474307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000474307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000474307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000474307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000474307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011129.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181582.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181582.hg.1 |
ENST00000367357.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181582.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181582.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181582.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181582.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181582.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181582.hg.1 |
ENST00000236914.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181582.hg.1 |
ENST00000367357.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181582.hg.1 |
ENST00000367362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181582.hg.1 |
ENST00000544748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100181582.hg.1 |
NM_001276464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181582.hg.1 |
NM_003822.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181582.hg.1 |
NM_205860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100181582.hg.1 |
TC0100011129.hg.1 |
Clariom D TC |