Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLFN12L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017024005.1 | XP_016879494.1 | 1 - 2 | 292 | 77 | |
XM_017024006.1 | XP_016879495.1 | 1 - 2 | 292 | 77 | ||
XM_017024007.1 | XP_016879496.1 | 1 - 2 | 292 | 77 | ||
XM_017024008.1 | XP_016879497.1 | 1 - 2 | 292 | 77 | ||
XM_017024009.1 | XP_016879498.1 | 1 - 2 | 292 | 77 | ||
GenBank mRNA | AK172761.1 | - | 1 - 2 | 127 | 77 | |
DA814276.1 | - | 1 - 2 | 292 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |