Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006689.hg.1 |
TC06000587.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006689.hg.1 |
ENST00000304672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006689.hg.1 |
ENST00000441198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006689.hg.1 |
ENST00000446507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006689.hg.1 |
NM_001243168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006689.hg.1 |
NM_001243169.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006689.hg.1 |
NM_001243170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006689.hg.1 |
NM_138296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
ENST00000304672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
ENST00000441198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
ENST00000446507.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
ENST00000616441.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
NM_001243168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
NM_001243169.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
NM_001243170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
NM_138296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
ENST00000304672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
ENST00000441198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
ENST00000446507.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
ENST00000616441.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
NM_001243168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
NM_001243169.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
NM_001243170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
NM_138296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155983.hg.1 |
TC0600008057.hg.1 |
Clariom D TC |