Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005907.hg.1 |
TC01000381.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005907.hg.1 |
ENST00000373831.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005907.hg.1 |
ENST00000373832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005907.hg.1 |
ENST00000373833.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005907.hg.1 |
ENST00000398958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005907.hg.1 |
ENST00000486790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005907.hg.1 |
NM_001048194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005907.hg.1 |
NM_001048195.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005907.hg.1 |
NM_001048199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005907.hg.1 |
NM_001269.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005907.hg.1 |
NR_030725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005907.hg.1 |
NR_030726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000373831.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000373832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000373833.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000398958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000411533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000419074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000427469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000430407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000434290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000486790.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
NM_001048194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
NM_001048195.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
NM_001048199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
NM_001269.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
NR_030725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
NR_030726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000373831.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000373832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000373833.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000398958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000411533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000419074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000427469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000430407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000434290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
ENST00000486790.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
NM_001048194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
NM_001048195.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
NM_001048199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
NM_001269.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
NR_030725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
NR_030726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152976.hg.1 |
TC0100007559.hg.1 |
Clariom D TC |