Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001860.hg.1 |
TC02004917.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001860.hg.1 |
ENST00000335756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001860.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221240.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221240.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221240.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221240.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221240.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221240.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221240.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221240.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221240.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221240.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221240.hg.1 |
TC0200016417.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000233505.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000335756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000419525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000460030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
ENST00000472719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001042426.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016417.hg.1 |
NM_001809.3 |
Refseq |