Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000427943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000427943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000427943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000427943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000427943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000427943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000497722.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000497722.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000497722.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000497722.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005141.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005141.hg.1 |
TC13000453.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005139.hg.1 |
ENST00000497722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005139.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005139.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005139.hg.1 |
TC13000453.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005142.hg.1 |
ENST00000423023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005142.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005142.hg.1 |
TC13000453.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005140.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005140.hg.1 |
TC13000453.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000338910.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000427943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000427943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000427943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000427943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000427943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000427943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000497722.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000497722.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000497722.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000497722.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
ENST00000619300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NM_001042414.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008225.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155547.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155547.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155547.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155547.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155547.hg.1 |
TC1300008225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300155546.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155546.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155546.hg.1 |
TC1300008225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300155548.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155548.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155548.hg.1 |
ENST00000492741.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155548.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155548.hg.1 |
TC1300008225.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300155545.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155545.hg.1 |
ENST00000497722.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155545.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155545.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155545.hg.1 |
ENST00000471658.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155545.hg.1 |
ENST00000497722.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155545.hg.1 |
NR_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155545.hg.1 |
NR_044998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155545.hg.1 |
TC1300008225.hg.1 |
Clariom D TC |