Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032961.2 | NP_116586.1 | 2 - 3 | 3539 | 76 | |
XM_011532150.1 | XP_011530452.1 | 2 - 3 | 3539 | 76 | ||
GenBank mRNA | AB037821.1 | - | 2 - 3 | 3514 | 76 | |
AK024636.1 | - | 2 - 3 | 803 | 76 | ||
AL157470.1 | - | 2 - 3 | 596 | 76 | ||
BC111560.1 | - | 2 - 3 | 2877 | 40068151 | 76 | |
KJ534913.1 | - | 2 - 3 | 2688 | 76 | ||
KJ534914.1 | - | 3 - 4 | 2517 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |