Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011020.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011020.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011020.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011020.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011020.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011020.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011020.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011020.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011020.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011020.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011020.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011020.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011020.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011020.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011020.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011020.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022209.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022209.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022209.hg.1 |
TC17001657.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022208.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022208.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022208.hg.1 |
TC17001657.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700188696.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188696.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188696.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188696.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188696.hg.1 |
TC1700011020.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700188698.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188698.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188698.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188698.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188698.hg.1 |
TC1700011020.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700011020.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011020.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011020.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011020.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011020.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011020.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011020.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011020.hg.1 |
ENST00000357424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011020.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011020.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011020.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011020.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011020.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011020.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011020.hg.1 |
NM_004123.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011020.hg.1 |
NM_004123.2 |
Refseq |