Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017749.hg.1 |
ENST00000372919.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017749.hg.1 |
ENST00000372921.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017749.hg.1 |
ENST00000535178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017749.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017749.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017749.hg.1 |
TC10001408.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000394847.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000394847.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000394847.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000394847.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000394847.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000394847.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000463788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000463788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000463788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000463788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000463788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175031.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175031.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175031.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175031.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175031.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175031.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175031.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175031.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175031.hg.1 |
TC1000011020.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372919.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000372921.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000394847.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000394847.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000394847.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000394847.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000394847.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000394847.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000463788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000463788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000463788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000463788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000463788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
ENST00000492380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_001156.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011020.hg.1 |
NM_004034.3 |
Refseq |