Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACRV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001612.5 | NP_001603.1 | 2 - 3 | 671 | 96 | |
NM_020107.4 | NP_064492.1 | 3 - 4 | 506 | 96 | ||
NM_020108.4 | NP_064493.1 | 3 - 4 | 461 | 96 | ||
GenBank mRNA | AK223335.1 | - | 2 - 3 | 620 | 96 | |
BC014588.2 | - | 2 - 3 | 647 | 4825974 | 96 | |
BG720361.1 | - | 3 - 4 | 431 | 96 | ||
BI826178.1 | - | 3 - 4 | 439 | 96 | ||
M82968.1 | - | 2 - 3 | 605 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |